The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014326.3 | NP_055141.2 | 3 - 4 | 347 | 87 |
XM_011521413.2 | XP_011519715.1 | 2 - 3 | 322 | 87 | |
XM_011521414.1 | XP_011519716.1 | 2 - 3 | 322 | 87 | |
XM_011521415.2 | XP_011519717.1 | 2 - 3 | 322 | 87 | |
XM_011521416.2 | XP_011519718.1 | 3 - 4 | 495 | 87 | |
XM_011521417.1 | XP_011519719.1 | 2 - 3 | 322 | 87 | |
XM_017022044.1 | XP_016877533.1 | 2 - 3 | 322 | 87 | |
XM_017022045.1 | XP_016877534.1 | 2 - 3 | 322 | 87 | |
XM_017022046.1 | XP_016877535.1 | 2 - 3 | 322 | 87 | |
GenBank mRNA | AB018001.1 | - | 3 - 4 | 346 | 87 |
AF052941.1 | - | 3 - 4 | 347 | 87 | |
AK310967.1 | - | 4 - 5 | 434 | 87 | |
BC114506.1 | - | 2 - 3 | 336 | 87 | |
GU056176.1 | - | 3 - 4 | 347 | 87 |