The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080860.3 | NP_543136.1 | 1 - 2 | 225 | 66 |
XM_011529786.1 | XP_011528088.1 | 1 - 2 | 117 | 66 | |
GenBank mRNA | AB006536.1 | - | 1 - 2 | 225 | 66 |
AB041016.1 | - | 1 - 2 | 162 | 66 | |
AK057315.1 | - | 1 - 2 | 117 | 66 | |
AK314746.1 | - | 1 - 2 | 62 | 66 | |
BC101519.1 | - | 1 - 2 | 104 | 66 | |
BC113367.1 | - | 1 - 2 | 104 | 66 |