Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
Chromosome Location
Chr.7: 122318424 - 122886759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009571.3 NP_001009571.2 2 - 3 865 72
NM_001167940.1 NP_001161412.1 2 - 3 865 72
NM_017954.10 NP_060424.9 2 - 3 865 72
XM_005250695.4 XP_005250752.1 2 - 3 594 72
XM_005250696.4 XP_005250753.1 2 - 3 594 72
XM_005250697.4 XP_005250754.1 2 - 3 594 72
XM_005250698.4 XP_005250755.1 2 - 3 566 72
XM_005250699.4 XP_005250756.1 2 - 3 594 72
XM_005250700.4 XP_005250757.1 2 - 3 594 72
XM_005250701.4 XP_005250758.1 2 - 3 594 72
XM_005250702.4 XP_005250759.1 2 - 3 594 72
XM_005250703.4 XP_005250760.1 2 - 3 594 72
XM_005250704.4 XP_005250761.1 2 - 3 594 72
XM_005250705.4 XP_005250762.1 2 - 3 594 72
XM_005250706.4 XP_005250763.1 2 - 3 594 72
XM_005250707.4 XP_005250764.1 2 - 3 594 72
XM_005250708.3 XP_005250765.1 2 - 3 594 72
XM_011516695.2 XP_011514997.1 2 - 3 566 72
XM_011516696.2 XP_011514998.1 2 - 3 594 72
XM_011516697.2 XP_011514999.1 2 - 3 594 72
XM_017012794.1 XP_016868283.1 2 - 3 594 72
XM_017012795.1 XP_016868284.1 2 - 3 594 72
XM_017012796.1 XP_016868285.1 3 - 4 209 72
XM_017012797.1 XP_016868286.1 3 - 4 275 72
XM_017012798.1 XP_016868287.1 2 - 3 745 72
XM_017012799.1 XP_016868288.1 3 - 4 646 72
GenBank mRNA AF401638.1 - 2 - 3 534 72
AK126518.1 - 3 - 4 467 72
AY264289.1 - 2 - 3 469 72
BC054339.1 - 2 - 3 543 72
BC136601.1 - 2 - 3 564 72
BC144278.1 - 2 - 3 564 72
BC144279.1 - 2 - 3 564 72
KJ534800.1 - 2 - 3 443 72
KJ534801.1 - 2 - 3 443 72
KJ534802.1 - 2 - 3 443 72
KJ534803.1 - 2 - 3 443 72
KJ534804.1 - 2 - 3 443 72
KJ534805.1 - 2 - 3 443 72
KJ535060.1 - 2 - 3 443 72

View Full Product Details