Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
Chromosome Location
Chr.7: 122318424 - 122886759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009571.3 NP_001009571.2 16 - 17 2894 119
NM_017954.10 NP_060424.9 17 - 18 2903 119
XM_005250699.4 XP_005250756.1 17 - 18 2632 119
XM_005250701.4 XP_005250758.1 17 - 18 2632 119
XM_005250702.4 XP_005250759.1 16 - 17 2623 119
XM_005250707.4 XP_005250764.1 17 - 18 2632 119
XM_005250708.3 XP_005250765.1 16 - 17 2623 119
XM_011516696.2 XP_011514998.1 17 - 18 2632 119
XM_011516697.2 XP_011514999.1 17 - 18 2632 119
XM_017012794.1 XP_016868283.1 16 - 17 2623 119
XM_017012798.1 XP_016868287.1 17 - 18 2783 119
GenBank mRNA AB046811.1 - 14 - 15 1763 119
AF401638.1 - 16 - 17 2563 119
AK000768.1 - 1 - 2 72 119
AK025672.1 - 12 - 13 1417 119
AK126518.1 - 17 - 18 2496 119
AL833058.1 - 13 - 14 1689 119
AY264289.1 - 17 - 18 2507 119
BC136601.1 - 16 - 17 2593 119
BC144278.1 - 16 - 17 2593 119
KJ534798.1 - 17 - 18 2481 119
KJ534800.1 - 16 - 17 2472 119
KJ534801.1 - 16 - 17 2472 119
KJ534802.1 - 17 - 18 2481 119
KJ534805.1 - 16 - 17 2472 119
KJ535060.1 - 4 - 5 707 119

View Full Product Details