Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SUOX
- Entrez Gene ID
-
6821
- Gene Name
- sulfite oxidase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 55996776 - 56005525 on Build GRCh38
- Gene Symbol
- RAB11B
- Entrez Gene ID
-
9230
- Gene Name
- RAB11B, member RAS oncogene family
- Gene Aliases
-
H-YPT3
- Chromosome Location
-
Chr.19: 8390321 - 8404434 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX647356.1 |
- |
- |
|
70 |
View Full Product Details