Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAN2
Entrez Gene ID
9924
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Gene Aliases
USP52
Chromosome Location
Chr.12: 56316223 - 56334053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127460.2 NP_001120932.1 12 - 13 2305 99
NM_014871.4 NP_055686.3 12 - 13 2234 99
GenBank mRNA AB107585.1 - 11 - 12 1928 99
BC024043.1 - 12 - 13 2147 99
BC094885.1 - 12 - 13 2302 99

View Full Product Details