The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323316.1 | NP_001310245.1 | 12 - 13 | 1991 | 72 |
NM_001323317.1 | NP_001310246.1 | 10 - 11 | 1765 | 72 | |
NM_001323318.1 | NP_001310247.1 | 10 - 11 | 1828 | 72 | |
NM_004613.3 | NP_004604.2 | 11 - 12 | 2008 | 72 | |
XM_011529028.1 | XP_011527330.1 | 12 - 13 | 1953 | 72 | |
GenBank mRNA | AK126508.1 | - | 1 - 2 | 1422 | 72 |
AK291714.1 | - | 11 - 12 | 1849 | 72 | |
AK295775.1 | - | 10 - 11 | 1606 | 72 | |
AK300292.1 | - | 10 - 11 | 1669 | 72 | |
AK314618.1 | - | 11 - 12 | 1849 | 72 | |
AY675221.1 | - | 10 - 11 | 1777 | 72 | |
BQ938957.1 | - | 3 - 4 | 569 | 72 | |
EU794665.1 | - | 11 - 12 | 1877 | 72 | |
M55153.1 | - | 11 - 12 | 1912 | 72 |