The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323316.1 | NP_001310245.1 | 6 - 7 | 893 | 63 |
NM_001323317.1 | NP_001310246.1 | 4 - 5 | 667 | 63 | |
NM_001323318.1 | NP_001310247.1 | 4 - 5 | 730 | 63 | |
NM_004613.3 | NP_004604.2 | 5 - 6 | 910 | 63 | |
NM_198951.2 | NP_945189.1 | 5 - 6 | 910 | 63 | |
XM_011529028.1 | XP_011527330.1 | 6 - 7 | 855 | 63 | |
GenBank mRNA | AK058031.1 | - | 5 - 6 | 678 | 63 |
AK291714.1 | - | 5 - 6 | 751 | 63 | |
AK295775.1 | - | 4 - 5 | 508 | 63 | |
AK300292.1 | - | 4 - 5 | 571 | 63 | |
AK314618.1 | - | 5 - 6 | 751 | 63 | |
AY675221.1 | - | 4 - 5 | 679 | 63 | |
BC003551.1 | - | 5 - 6 | 763 | 63 | |
EU794665.1 | - | 5 - 6 | 779 | 63 | |
S81734.1 | - | 5 - 6 | 811 | 63 |