Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS8
Entrez Gene ID
23355
Gene Name
VPS8, CORVET complex subunit
Gene Aliases
KIAA0804
Chromosome Location
Chr.3: 184812137 - 185052614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009921.2 NP_001009921.1 2 - 3 321 147
NM_015303.3 NP_056118.2 2 - 3 321 147
XM_005247251.3 XP_005247308.1 2 - 3 589 147
XM_005247253.4 XP_005247310.1 2 - 3 327 147
XM_017006041.1 XP_016861530.1 2 - 3 327 147
GenBank mRNA AK056661.1 - 2 - 3 293 147
AK292423.1 - 2 - 3 293 147
BC140768.1 - 2 - 3 257 147
BX647915.1 - 2 - 3 283 147
DA218768.1 - 2 - 3 328 147
DA340096.1 - 2 - 3 278 147

View Full Product Details