Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS8
Entrez Gene ID
23355
Gene Name
VPS8, CORVET complex subunit
Gene Aliases
KIAA0804
Chromosome Location
Chr.3: 184812137 - 185052614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009921.2 NP_001009921.1 4 - 5 528 86
NM_015303.3 NP_056118.2 4 - 5 528 86
XM_005247251.3 XP_005247308.1 4 - 5 796 86
XM_005247253.4 XP_005247310.1 4 - 5 534 86
XM_017006040.1 XP_016861529.1 3 - 4 222 86
XM_017006041.1 XP_016861530.1 4 - 5 534 86
GenBank mRNA AK056661.1 - 4 - 5 500 86
AK292423.1 - 4 - 5 500 86
AL833838.1 - 1 - 2 61 86
BC140768.1 - 4 - 5 464 86
BX647915.1 - 4 - 5 490 86
DA340096.1 - 4 - 5 485 86

View Full Product Details