Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXD2
Entrez Gene ID
55218
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
C14orf114, EXDL2
Chromosome Location
Chr.14: 69191477 - 69244020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193361.1 NP_001180290.1 1 - 2 115 105
GenBank mRNA BX647767.1 - 1 - 2 64 105
CR936729.1 - 1 - 2 82 105

View Full Product Details