Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA1
Entrez Gene ID
5265
Gene Name
serpin family A member 1
Gene Aliases
A1A, A1AT, AAT, PI, PI1, PRO2275, alpha1AT
Chromosome Location
Chr.14: 94376747 - 94390692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017021370.1 XP_016876859.1 2 - 3 304 85

View Full Product Details