Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
Chromosome Location
Chr.6: 75921114 - 76072678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282368.1 NP_001269297.1 14 - 15 2200 176
NM_001563.3 NP_001554.2 15 - 16 2434 176
GenBank mRNA AF047492.2 - 15 - 16 2374 176
BC117450.1 - 15 - 16 2264 176
BC117452.1 - 15 - 16 2264 176
CR749572.1 - 15 - 16 4676 176

View Full Product Details