The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256088.1 | NP_001243017.1 | 2 - 3 | 337 | 76 |
NM_145013.2 | NP_659450.1 | 2 - 3 | 326 | 76 | |
NR_045767.1 | 2 - 3 | 260 | 76 | ||
XM_011542654.2 | XP_011540956.1 | 2 - 3 | 256 | 76 | |
XM_011542655.2 | XP_011540957.1 | 2 - 3 | 218 | 76 | |
GenBank mRNA | AK125634.1 | - | 2 - 3 | 234 | 76 |
AK314137.1 | - | 2 - 3 | 303 | 76 | |
BC025756.1 | - | 2 - 3 | 326 | 76 |