The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276506.1 | NP_001263435.1 | 2 - 3 | 253 | 123 |
NM_003002.3 | NP_002993.1 | 2 - 3 | 253 | 123 | |
NR_077060.1 | 2 - 3 | 253 | 123 | ||
GenBank mRNA | AB006202.1 | - | 2 - 3 | 180 | 123 |
AK075360.1 | - | 2 - 3 | 179 | 123 | |
BC005263.1 | - | 2 - 3 | 197 | 123 | |
BC009574.1 | - | 2 - 3 | 178 | 123 | |
BC012603.1 | - | 2 - 3 | 182 | 123 | |
BC015188.1 | - | 2 - 3 | 204 | 123 | |
BC015992.1 | - | 2 - 3 | 200 | 123 | |
BC022350.1 | - | 2 - 3 | 192 | 123 | |
BC071755.1 | - | 2 - 3 | 196 | 123 | |
BC071756.1 | - | 2 - 3 | 178 | 123 | |
BG492859.1 | - | 2 - 3 | 180 | 123 | |
BQ932668.1 | - | 2 - 3 | 261 | 123 | |
BT007238.1 | - | 2 - 3 | 169 | 123 | |
BX419391.2 | - | 2 - 3 | 230 | 123 | |
CR456932.1 | - | 2 - 3 | 169 | 123 | |
KU178325.1 | - | 2 - 3 | 169 | 123 |