The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014427.4 | NP_055242.1 | 14 - 15 | 1524 | 73 |
NM_153636.2 | NP_705900.1 | 12 - 13 | 1299 | 73 | |
XM_011523000.1 | XP_011521302.1 | 12 - 13 | 1294 | 73 | |
XM_011523001.2 | XP_011521303.1 | 12 - 13 | 1059 | 73 | |
XM_017023138.1 | XP_016878627.1 | 14 - 15 | 1488 | 73 | |
XM_017023139.1 | XP_016878628.1 | 14 - 15 | 1499 | 73 | |
XM_017023140.1 | XP_016878629.1 | 12 - 13 | 1274 | 73 | |
XM_017023141.1 | XP_016878630.1 | 11 - 12 | 2381 | 73 | |
GenBank mRNA | AJ133798.1 | - | 14 - 15 | 1523 | 73 |
BC035334.1 | - | 12 - 13 | 1237 | 73 | |
BC064577.1 | - | 12 - 13 | 1276 | 73 |