The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109662.3 | NP_001103132.3 | 15 - 16 | 2464 | 112 |
GenBank mRNA | AB777655.1 | - | 15 - 16 | 2816 | 112 |
AK091473.1 | - | 5 - 6 | 657 | 112 | |
BC065706.1 | - | 5 - 6 | 698 | 112 | |
BC101482.1 | - | 4 - 5 | 573 | 112 | |
BC143384.1 | - | 4 - 5 | 573 | 112 | |
BC143388.1 | - | 4 - 5 | 573 | 112 |