Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
Chromosome Location
Chr.16: 89575768 - 89597246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014427.4 NP_055242.1 4 - 5 714 141
XM_017023138.1 XP_016878627.1 4 - 5 678 141
XM_017023139.1 XP_016878628.1 4 - 5 689 141
GenBank mRNA AJ133798.1 - 4 - 5 713 141

View Full Product Details