The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002690.2 | NP_002681.1 | 10 - 11 | 793 | 64 |
XM_005273535.3 | XP_005273592.1 | 11 - 12 | 895 | 64 | |
XM_005273538.2 | XP_005273595.1 | 10 - 11 | 805 | 64 | |
XM_005273539.2 | XP_005273596.1 | 9 - 10 | 700 | 64 | |
XM_005273540.4 | XP_005273597.1 | 6 - 7 | 554 | 64 | |
XM_006716353.2 | XP_006716416.1 | 7 - 8 | 439 | 64 | |
XM_017013583.1 | XP_016869072.1 | 8 - 9 | 697 | 64 | |
XM_017013584.1 | XP_016869073.1 | 7 - 8 | 590 | 64 | |
GenBank mRNA | AK223537.1 | - | 10 - 11 | 792 | 64 |
AK294025.1 | - | 6 - 7 | 512 | 64 | |
AK314976.1 | - | 10 - 11 | 754 | 64 | |
BC100288.1 | - | 10 - 11 | 757 | 64 | |
BC106909.1 | - | 10 - 11 | 719 | 64 | |
CR536503.1 | - | 10 - 11 | 623 | 64 | |
CR541802.1 | - | 10 - 11 | 623 | 64 | |
D29013.1 | - | 10 - 11 | 736 | 64 | |
L11607.1 | - | 10 - 11 | 623 | 64 | |
M13140.1 | - | 10 - 11 | 736 | 64 |