The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040260.3 | NP_001035350.2 | 7 - 8 | 1996 | 100 |
NM_001040261.4 | NP_001035351.4 | 8 - 9 | 2047 | 100 | |
NR_036614.1 | 7 - 8 | 1993 | 100 | ||
XM_005262785.2 | XP_005262842.1 | 8 - 9 | 1873 | 100 | |
XM_005262786.2 | XP_005262843.1 | 7 - 8 | 1822 | 100 | |
XM_017007832.1 | XP_016863321.1 | 8 - 9 | 1876 | 100 | |
GenBank mRNA | AB209181.1 | - | 7 - 8 | 1996 | 100 |
AL834498.1 | - | 8 - 9 | 1094 | 100 | |
BC032726.1 | - | 7 - 8 | 1286 | 100 | |
DA201927.1 | - | 5 - 6 | 443 | 100 |