Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGF
Entrez Gene ID
1950
Gene Name
epidermal growth factor
Gene Aliases
HOMG4, URG
Chromosome Location
Chr.4: 109912883 - 110013079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178130.2 NP_001171601.1 7 - 8 1638 72
NM_001178131.2 NP_001171602.1 6 - 7 1512 72
NM_001963.5 NP_001954.2 7 - 8 1638 72
XM_005262796.2 XP_005262853.1 7 - 8 1639 72
XM_011531707.1 XP_011530009.1 8 - 9 1288 72
XM_017007845.1 XP_016863334.1 7 - 8 1663 72
XM_017007846.1 XP_016863335.1 7 - 8 1663 72
XM_017007847.1 XP_016863336.1 7 - 8 1663 72
XM_017007848.1 XP_016863337.1 6 - 7 1537 72
XM_017007849.1 XP_016863338.1 7 - 8 1663 72
XM_017007850.1 XP_016863339.1 7 - 8 1663 72
XM_017007851.1 XP_016863340.1 7 - 8 1663 72
XM_017007852.1 XP_016863341.1 6 - 7 1513 72
XM_017007853.1 XP_016863342.1 7 - 8 1663 72
XM_017007854.1 XP_016863343.1 7 - 8 1663 72
XM_017007855.1 XP_016863344.1 7 - 8 1663 72
GenBank mRNA AK299306.1 - 6 - 7 1514 72
BC093731.1 - 7 - 8 1281 72
BC113461.1 - 7 - 8 1281 72
BC143357.1 - 7 - 8 1281 72
X04571.1 - 7 - 8 1622 72

View Full Product Details