The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003363.3 | NP_003354.2 | 14 - 15 | 1962 | 96 |
NM_199443.2 | NP_955475.1 | 13 - 14 | 1821 | 96 | |
GenBank mRNA | AF017305.1 | - | 14 - 15 | 1886 | 96 |
AF017306.1 | - | 13 - 14 | 1742 | 96 | |
AK127055.1 | - | 8 - 9 | 2398 | 96 | |
AK222725.1 | - | 13 - 14 | 1751 | 96 | |
AK223292.1 | - | 14 - 15 | 1892 | 96 | |
AK291795.1 | - | 14 - 15 | 1907 | 96 | |
BC125130.1 | - | 14 - 15 | 1933 | 96 | |
BC125131.1 | - | 14 - 15 | 1930 | 96 | |
U20657.1 | - | 14 - 15 | 1886 | 96 |