Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
Chromosome Location
Chr.11: 95790461 - 95832693 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243776.1 NP_001230705.1 4 - 5 664 158
NM_001243777.1 NP_001230706.1 3 - 4 620 158
NM_014679.4 NP_055494.2 3 - 4 620 158
XM_006718945.2 XP_006719008.1 3 - 4 620 158
XM_006718946.2 XP_006719009.1 3 - 4 620 158
XM_017018592.1 XP_016874081.1 4 - 5 1810 158
XM_017018593.1 XP_016874082.1 3 - 4 620 158
XM_017018594.1 XP_016874083.1 3 - 4 620 158
GenBank mRNA AK098647.1 - 3 - 4 593 158
AK291245.1 - 3 - 4 575 158
AK293277.1 - 4 - 5 617 158
AY225092.1 - 3 - 4 382 158
AY239292.1 - 3 - 4 521 158
BC001233.2 - 3 - 4 603 158
BC029385.1 - 3 - 4 386 158
BC031048.1 - 3 - 4 531 158
BC039711.1 - 3 - 4 429 158
D42054.1 - 3 - 4 435 158

View Full Product Details