The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127396.2 | NP_001120868.1 | 5 - 6 | 367 | 66 |
NM_001272034.1 | NP_001258963.1 | 5 - 6 | 409 | 66 | |
NM_006949.3 | NP_008880.2 | 5 - 6 | 376 | 66 | |
NR_073560.1 | 5 - 6 | 376 | 66 | ||
XM_011528210.1 | XP_011526512.1 | 5 - 6 | 372 | 66 | |
XM_011528212.2 | XP_011526514.1 | 5 - 6 | 372 | 66 | |
GenBank mRNA | AB002559.1 | - | 5 - 6 | 351 | 66 |
AF318317.1 | - | 1 - 2 | 1547 | 66 | |
AK222967.1 | - | 5 - 6 | 353 | 66 | |
AK222977.1 | - | 5 - 6 | 353 | 66 | |
AK302257.1 | - | 5 - 6 | 331 | 66 | |
AK303701.1 | - | 5 - 6 | 397 | 66 | |
AK314230.1 | - | 5 - 6 | 355 | 66 | |
BC002869.2 | - | 5 - 6 | 341 | 66 | |
BQ067493.1 | - | 5 - 6 | 377 | 66 | |
BT006915.1 | - | 5 - 6 | 318 | 66 | |
DA431480.1 | - | 5 - 6 | 372 | 66 | |
U63533.1 | - | 5 - 6 | 351 | 66 |