The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243532.1 | NP_001230461.1 | 2 - 3 | 564 | 78 |
NM_001243533.1 | NP_001230462.1 | 3 - 4 | 638 | 78 | |
NM_152835.4 | NP_690048.1 | 2 - 3 | 447 | 78 | |
XM_005245743.4 | XP_005245800.1 | 3 - 4 | 707 | 78 | |
GenBank mRNA | AF411102.1 | - | 3 - 4 | 578 | 78 |
AK312872.1 | - | 2 - 3 | 447 | 78 | |
BC028713.1 | - | 3 - 4 | 580 | 78 | |
CV024336.1 | - | 1 - 2 | 291 | 78 | |
DB227864.1 | - | 2 - 3 | 447 | 78 |