Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FUNDC2P2
- Entrez Gene ID
-
388965
- Gene Name
- FUN14 domain containing 2 pseudogene 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 84290682 - 84292200 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003663.1 |
|
1 - 1 |
1101 |
99 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AW592923.1 |
- |
3 |
|
|
Chr. 2 - 84291644 - 84303638 [-] on Build GRCh38 |
100 |
GenBankmRNA |
DB539553.2 |
- |
1 |
432 |
|
Chr. 2 - 84291352 - 84303641 [+] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928031 |
LOC101928031 |
|
-767.9kb
|
Sense |
Chr. - 83522814 - 83523482 on Build GRCh38 |
Downstream |
succinate-CoA ligase alpha subunit |
SUCLG1 |
GALPHA, MTDPS9, SUCLA1 |
131.7kb
|
Anti-sense |
Chr. - 84423523 - 84459462 on Build GRCh38 |
View Full Product Details