The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206671.2 | NP_001193600.1 | 5 - 6 | 729 | 82 |
NM_024557.4 | NP_078833.3 | 5 - 6 | 726 | 82 | |
NR_045405.1 | 6 - 7 | 778 | 82 | ||
XM_006718317.3 | XP_006718380.1 | 5 - 6 | 858 | 82 | |
XM_006718318.3 | XP_006718381.1 | 5 - 6 | 771 | 82 | |
XM_017018287.1 | XP_016873776.1 | 5 - 6 | 858 | 82 | |
XM_017018288.1 | XP_016873777.1 | 6 - 7 | 855 | 82 | |
XM_017018289.1 | XP_016873778.1 | 6 - 7 | 802 | 82 | |
GenBank mRNA | AK021670.1 | - | 4 - 5 | 750 | 82 |
AK293933.1 | - | 5 - 6 | 776 | 82 | |
AK296505.1 | - | 6 - 7 | 743 | 82 | |
AK315379.1 | - | 5 - 6 | 685 | 82 | |
AY326435.1 | - | 5 - 6 | 675 | 82 | |
BC022455.1 | - | 5 - 6 | 708 | 82 | |
KU178773.1 | - | 5 - 6 | 672 | 82 | |
KU178774.1 | - | 4 - 5 | 578 | 82 |