Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr.16: 81100879 - 81220370 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278425.1 NP_001265354.1 23 - 24 3952 65
NM_052892.3 NP_443124.3 34 - 35 5858 65
NR_126532.1 34 - 35 5858 65
GenBank mRNA AB067466.1 - 25 - 26 5271 65
AK131378.1 - 2 - 3 247 65
AY164483.1 - 34 - 35 5858 65
AY371495.1 - 23 - 24 3914 65
BC014120.2 - 3 - 4 439 65

View Full Product Details