The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278425.1 | NP_001265354.1 | 11 - 12 | 1790 | 111 |
NM_052892.3 | NP_443124.3 | 22 - 23 | 3696 | 111 | |
NR_126532.1 | 22 - 23 | 3696 | 111 | ||
GenBank mRNA | AB067466.1 | - | 13 - 14 | 3109 | 111 |
AY164483.1 | - | 22 - 23 | 3696 | 111 | |
AY371495.1 | - | 11 - 12 | 1752 | 111 |