The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001975.2 | NP_001966.1 | 11 - 12 | 1451 | 83 |
GenBank mRNA | AK124656.1 | - | 10 - 11 | 1855 | 83 |
AK290525.1 | - | 11 - 12 | 1577 | 83 | |
AK295220.1 | - | 9 - 10 | 1183 | 83 | |
AK312402.1 | - | 11 - 12 | 1318 | 83 | |
BC002745.2 | - | 11 - 12 | 1305 | 83 | |
BT007383.1 | - | 10 - 11 | 1229 | 83 | |
CR536582.1 | - | 10 - 11 | 1229 | 83 | |
EU794607.1 | - | 10 - 11 | 1229 | 83 | |
M22349.1 | - | 11 - 12 | 1303 | 83 | |
M36768.1 | - | 9 - 10 | 1151 | 83 |