Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
Chromosome Location
Chr.12: 121308245 - 121354209 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137559.1 NP_001131031.1 2 - 3 180 58
NM_016237.4 NP_057321.2 2 - 3 402 58
XM_005253900.3 XP_005253957.1 2 - 3 404 58
XM_017019423.1 XP_016874912.1 2 - 3 120 58
XM_017019424.1 XP_016874913.1 2 - 3 120 58
GenBank mRNA AF191339.1 - 2 - 3 280 58
AI879332.1 - 2 - 3 404 58
AK025614.1 - 2 - 3 180 58
AK315451.1 - 2 - 3 348 58
BC001081.2 - 2 - 3 335 58
BC001950.1 - 2 - 3 335 58
BC006301.2 - 2 - 3 339 58
BC034243.1 - 1 - 2 42 58

View Full Product Details