The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001137559.1 | NP_001131031.1 | 5 - 6 | 559 | 89 |
NM_016237.4 | NP_057321.2 | 5 - 6 | 781 | 89 | |
XM_005253900.3 | XP_005253957.1 | 5 - 6 | 783 | 89 | |
XM_017019423.1 | XP_016874912.1 | 5 - 6 | 499 | 89 | |
XM_017019424.1 | XP_016874913.1 | 5 - 6 | 499 | 89 | |
GenBank mRNA | AF191339.1 | - | 5 - 6 | 659 | 89 |
AK025614.1 | - | 5 - 6 | 559 | 89 | |
AK315451.1 | - | 5 - 6 | 726 | 89 | |
BC001081.2 | - | 5 - 6 | 714 | 89 | |
BC001950.1 | - | 5 - 6 | 714 | 89 | |
BC006301.2 | - | 5 - 6 | 718 | 89 | |
BC034243.1 | - | 4 - 5 | 421 | 89 | |
BX537687.1 | - | 2 - 3 | 462 | 89 |