The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174937.3 | NP_777597.2 | 10 - 11 | 1568 | 122 |
XM_011539577.1 | XP_011537879.1 | 11 - 12 | 1601 | 122 | |
GenBank mRNA | BC042951.1 | - | 10 - 11 | 1573 | 122 |
BC093639.1 | - | 10 - 11 | 1417 | 122 | |
BC101535.1 | - | 10 - 11 | 1417 | 122 |