The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024592.4 | NP_078868.1 | 4 - 5 | 818 | 76 |
XM_017008601.1 | XP_016864090.1 | 4 - 5 | 593 | 76 | |
GenBank mRNA | AK023414.1 | - | 4 - 5 | 798 | 76 |
BC002480.2 | - | 4 - 5 | 794 | 76 | |
CR457312.1 | - | 4 - 5 | 702 | 76 |