The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288715.1 | NP_001275644.1 | 16 - 17 | 3001 | 63 |
NM_001288716.1 | NP_001275645.1 | 14 - 15 | 2263 | 63 | |
NM_001288717.1 | NP_001275646.1 | 16 - 17 | 2628 | 63 | |
NM_001332.3 | NP_001323.1 | 17 - 18 | 3193 | 63 | |
NR_109988.1 | 16 - 17 | 2530 | 63 | ||
XM_005248251.3 | XP_005248308.1 | 18 - 19 | 3292 | 63 | |
XM_005248252.2 | XP_005248309.1 | 18 - 19 | 3458 | 63 | |
XM_005248253.1 | XP_005248310.1 | 17 - 18 | 2983 | 63 | |
XM_011513967.2 | XP_011512269.1 | 17 - 18 | 3372 | 63 | |
XM_017009072.1 | XP_016864561.1 | 16 - 17 | 2542 | 63 | |
XM_017009073.1 | XP_016864562.1 | 16 - 17 | 2717 | 63 | |
XM_017009074.1 | XP_016864563.1 | 15 - 16 | 2464 | 63 | |
XM_017009075.1 | XP_016864564.1 | 15 - 16 | 2591 | 63 | |
XM_017009076.1 | XP_016864565.1 | 15 - 16 | 2602 | 63 | |
GenBank mRNA | AB013805.1 | - | 17 - 18 | 3149 | 63 |
AF035302.1 | - | 5 - 6 | 770 | 63 | |
AK127257.1 | - | 11 - 12 | 2088 | 63 | |
AK226124.1 | - | 16 - 17 | 2880 | 63 | |
AK294420.1 | - | 14 - 15 | 2056 | 63 | |
AK296232.1 | - | 16 - 17 | 2628 | 63 | |
AK299300.1 | - | 14 - 15 | 2194 | 63 | |
EU328348.1 | - | 16 - 17 | 2467 | 63 | |
U52351.1 | - | 11 - 12 | 1958 | 63 | |
U52828.1 | - | 3 - 4 | 411 | 63 | |
U72665.1 | - | 6 - 7 | 940 | 63 | |
U81004.1 | - | 12 - 13 | 2449 | 63 | |
U96136.1 | - | 17 - 18 | 3003 | 63 |