Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
Chromosome Location
Chr.12: 75275979 - 75390941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286547.1 NP_001273476.1 14 - 15 1520 118
NM_001286548.1 NP_001273477.1 16 - 17 1893 118
NM_001286549.1 NP_001273478.1 9 - 10 1068 118
NM_032606.3 NP_115995.2 16 - 17 1765 118
XM_005269192.1 XP_005269249.1 14 - 15 1444 118
XM_005269194.3 XP_005269251.1 9 - 10 975 118
XM_006719648.2 XP_006719711.2 16 - 17 1765 118
XM_006719649.2 XP_006719712.2 15 - 16 1669 118
XM_011538878.1 XP_011537180.1 16 - 17 1730 118
XM_011538886.2 XP_011537188.1 15 - 16 1699 118
XM_011538887.2 XP_011537189.1 14 - 15 1603 118
XM_011538888.1 XP_011537190.1 12 - 13 1284 118
XM_011538889.2 XP_011537191.1 16 - 17 1636 118
XM_011538892.1 XP_011537194.1 13 - 14 1330 118
XM_017020034.1 XP_016875523.1 16 - 17 1592 118
XM_017020035.1 XP_016875524.1 11 - 12 1188 118
XM_017020036.1 XP_016875525.1 17 - 18 1855 118
XM_017020037.1 XP_016875526.1 14 - 15 1484 118
XM_017020038.1 XP_016875527.1 12 - 13 1352 118
XM_017020039.1 XP_016875528.1 10 - 11 1052 118
XM_017020040.1 XP_016875529.1 5 - 6 624 118
XM_017020041.1 XP_016875530.1 14 - 15 1590 118
XM_017020042.1 XP_016875531.1 14 - 15 1618 118
XM_017020043.1 XP_016875532.1 10 - 11 1435 118
XM_017020044.1 XP_016875533.1 9 - 10 1029 118
XM_017020045.1 XP_016875534.1 10 - 11 1471 118
XM_017020046.1 XP_016875535.1 9 - 10 1028 118
XM_017020047.1 XP_016875536.1 8 - 9 4481 118
GenBank mRNA AF251056.1 - 16 - 17 1893 118
AF351611.1 - 15 - 16 1474 118
AK091839.1 - 16 - 17 1765 118
AK092274.1 - 10 - 11 981 118
BC033860.1 - 14 - 15 1489 118
BC056672.1 - 10 - 11 1045 118
BC131724.1 - 9 - 10 1067 118

View Full Product Details