Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPS2
Entrez Gene ID
84698
Gene Name
calcyphosine 2
Gene Aliases
UG0636c06
Chromosome Location
Chr.12: 75275979 - 75390941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286548.1 NP_001273477.1 17 - 18 1996 145
NM_001286549.1 NP_001273478.1 10 - 11 1171 145
XM_005269192.1 XP_005269249.1 15 - 16 1547 145
XM_006719648.2 XP_006719711.2 17 - 18 1868 145
XM_006719649.2 XP_006719712.2 16 - 17 1772 145
XM_011538878.1 XP_011537180.1 17 - 18 1833 145
XM_011538886.2 XP_011537188.1 16 - 17 1802 145
XM_011538887.2 XP_011537189.1 15 - 16 1706 145
XM_011538888.1 XP_011537190.1 13 - 14 1387 145
XM_011538889.2 XP_011537191.1 17 - 18 1739 145
XM_011538892.1 XP_011537194.1 14 - 15 1433 145
XM_017020034.1 XP_016875523.1 17 - 18 1695 145
XM_017020035.1 XP_016875524.1 12 - 13 1291 145
XM_017020036.1 XP_016875525.1 18 - 19 1958 145
XM_017020037.1 XP_016875526.1 15 - 16 1587 145
XM_017020038.1 XP_016875527.1 13 - 14 1455 145
XM_017020039.1 XP_016875528.1 11 - 12 1155 145
XM_017020040.1 XP_016875529.1 6 - 7 727 145
XM_017020041.1 XP_016875530.1 15 - 16 1693 145
XM_017020042.1 XP_016875531.1 15 - 16 1721 145
XM_017020043.1 XP_016875532.1 11 - 12 1538 145
XM_017020044.1 XP_016875533.1 10 - 11 1132 145
XM_017020045.1 XP_016875534.1 11 - 12 1574 145
XM_017020046.1 XP_016875535.1 10 - 11 1131 145
XM_017020047.1 XP_016875536.1 9 - 10 4584 145
GenBank mRNA AF251056.1 - 17 - 18 1996 145
AF351611.1 - 16 - 17 1577 145
AK092274.1 - 11 - 12 1084 145
BC131724.1 - 10 - 11 1170 145

View Full Product Details