The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024630.2 | NP_078906.2 | 7 - 8 | 1463 | 55 |
NM_153746.1 | NP_714968.1 | 7 - 8 | 1463 | 55 | |
XM_005267146.3 | XP_005267203.1 | 6 - 7 | 834 | 55 | |
XM_011536120.2 | XP_011534422.1 | 7 - 8 | 2044 | 55 | |
XM_011536123.2 | XP_011534425.1 | 7 - 8 | 855 | 55 | |
XM_017011307.1 | XP_016866796.1 | 7 - 8 | 1148 | 55 | |
XM_017011308.1 | XP_016866797.1 | 7 - 8 | 1148 | 55 | |
XM_017011309.1 | XP_016866798.1 | 7 - 8 | 1148 | 55 | |
XM_017011310.1 | XP_016866799.1 | 7 - 8 | 1148 | 55 | |
XM_017011311.1 | XP_016866800.1 | 6 - 7 | 1010 | 55 | |
XM_017011312.1 | XP_016866801.1 | 7 - 8 | 1163 | 55 | |
XM_017011313.1 | XP_016866802.1 | 8 - 9 | 3860 | 55 | |
GenBank mRNA | AF542388.1 | - | 7 - 8 | 1463 | 55 |
AF542389.1 | - | 7 - 8 | 1463 | 55 | |
AK024637.1 | - | 3 - 4 | 272 | 55 | |
AK092380.1 | - | 3 - 4 | 1974 | 55 | |
AK124573.1 | - | 4 - 5 | 3830 | 55 | |
AK128773.1 | - | 3 - 4 | 2782 | 55 | |
BC029881.1 | - | 6 - 7 | 577 | 55 | |
BC056399.1 | - | 1 - 2 | 105 | 55 | |
BC125072.1 | - | 8 - 9 | 980 | 55 | |
BC125073.1 | - | 8 - 9 | 980 | 55 |