The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135642.1 | NP_001129114.1 | 12 - 13 | 1970 | 113 |
NM_016532.3 | NP_057616.2 | 10 - 11 | 1580 | 113 | |
NM_130766.2 | NP_570122.1 | 11 - 12 | 1811 | 113 | |
XM_005256683.2 | XP_005256740.1 | 10 - 11 | 1420 | 113 | |
XM_005256685.1 | XP_005256742.1 | 9 - 10 | 1201 | 113 | |
XM_005256686.1 | XP_005256743.1 | 10 - 11 | 1549 | 113 | |
XM_011523934.1 | XP_011522236.1 | 12 - 13 | 1593 | 113 | |
XM_011523936.2 | XP_011522238.1 | 10 - 11 | 1430 | 113 | |
XM_017024755.1 | XP_016880244.1 | 11 - 12 | 1651 | 113 | |
XM_017024756.1 | XP_016880245.1 | 12 - 13 | 1728 | 113 | |
XM_017024757.1 | XP_016880246.1 | 11 - 12 | 1626 | 113 | |
XM_017024758.1 | XP_016880247.1 | 10 - 11 | 1360 | 113 | |
XM_017024759.1 | XP_016880248.1 | 9 - 10 | 1211 | 113 | |
GenBank mRNA | AB036829.1 | - | 10 - 11 | 1365 | 113 |
AB036830.1 | - | 11 - 12 | 1594 | 113 | |
AK297955.1 | - | 8 - 9 | 1095 | 113 | |
AK312585.1 | - | 10 - 11 | 1365 | 113 | |
AK312844.1 | - | 10 - 11 | 1580 | 113 | |
BC004362.1 | - | 10 - 11 | 1379 | 113 | |
U45973.1 | - | 7 - 8 | 834 | 113 |