The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207307.2 | NP_997190.1 | 6 - 7 | 1410 | 77 |
XM_011513293.2 | XP_011511595.1 | 6 - 7 | 3484 | 77 | |
XM_011513294.2 | XP_011511596.1 | 6 - 7 | 3482 | 77 | |
XM_011513298.1 | XP_011511600.1 | 3 - 4 | 915 | 77 | |
XM_011513299.2 | XP_011511601.1 | 3 - 4 | 1109 | 77 | |
GenBank mRNA | AK096099.1 | - | 4 - 5 | 836 | 77 |
AL133011.1 | - | 4 - 5 | 4864 | 77 | |
BC066975.1 | - | 6 - 7 | 1410 | 77 |