Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EPB41L1
- Entrez Gene ID
-
2036
- Gene Name
- erythrocyte membrane protein band 4.1 like 1
- Gene Aliases
-
4.1N, MRD11
- Chromosome Location
-
Chr.20: 36064891 - 36232799 on Build GRCh38
View Full Product Details