The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258329.1 | NP_001245258.1 | 13 - 14 | 1674 | 58 |
NM_012156.2 | NP_036288.2 | 12 - 13 | 1620 | 58 | |
XM_011528667.1 | XP_011526969.1 | 17 - 18 | 2566 | 58 | |
XM_011528671.1 | XP_011526973.1 | 13 - 14 | 1653 | 58 | |
XM_011528673.1 | XP_011526975.1 | 12 - 13 | 1613 | 58 | |
XM_011528677.1 | XP_011526979.1 | 12 - 13 | 1427 | 58 | |
XM_011528683.1 | XP_011526985.1 | 11 - 12 | 1463 | 58 | |
XM_011528685.1 | XP_011526987.1 | 11 - 12 | 1463 | 58 | |
XM_017027711.1 | XP_016883200.1 | 11 - 12 | 1376 | 58 | |
XM_017027712.1 | XP_016883201.1 | 12 - 13 | 1514 | 58 | |
XM_017027716.1 | XP_016883205.1 | 13 - 14 | 1653 | 58 | |
XM_017027717.1 | XP_016883206.1 | 12 - 13 | 1611 | 58 | |
XM_017027718.1 | XP_016883207.1 | 12 - 13 | 1490 | 58 | |
GenBank mRNA | AB002336.1 | - | 12 - 13 | 1610 | 58 |
AK299817.1 | - | 12 - 13 | 1567 | 58 | |
AY049789.1 | - | 11 - 12 | 1458 | 58 | |
BC040259.1 | - | 13 - 14 | 1652 | 58 |