Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41L1
Entrez Gene ID
2036
Gene Name
erythrocyte membrane protein band 4.1 like 1
Gene Aliases
4.1N, MRD11
Chromosome Location
Chr.20: 36064891 - 36232799 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258329.1 NP_001245258.1 22 - 23 2860 64
NM_001258330.1 NP_001245259.1 20 - 21 2278 64
NM_001258331.1 NP_001245260.1 19 - 20 2505 64
NM_012156.2 NP_036288.2 21 - 22 2809 64
NM_177996.2 NP_818932.1 19 - 20 2500 64
XM_011528667.1 XP_011526969.1 28 - 29 5792 64
XM_011528671.1 XP_011526973.1 24 - 25 4879 64
XM_011528673.1 XP_011526975.1 23 - 24 4839 64
XM_011528677.1 XP_011526979.1 23 - 24 4653 64
XM_011528681.1 XP_011526983.1 19 - 20 2613 64
XM_011528682.1 XP_011526984.1 19 - 20 2610 64
XM_011528683.1 XP_011526985.1 19 - 20 2568 64
XM_011528685.1 XP_011526987.1 19 - 20 2241 64
XM_017027711.1 XP_016883200.1 22 - 23 4602 64
XM_017027712.1 XP_016883201.1 21 - 22 2703 64
XM_017027713.1 XP_016883202.1 20 - 21 2646 64
XM_017027714.1 XP_016883203.1 19 - 20 2559 64
XM_017027715.1 XP_016883204.1 19 - 20 2613 64
XM_017027716.1 XP_016883205.1 22 - 23 2839 64
XM_017027717.1 XP_016883206.1 21 - 22 2797 64
XM_017027718.1 XP_016883207.1 20 - 21 2268 64
XM_017027719.1 XP_016883208.1 19 - 20 2207 64
XM_017027720.1 XP_016883209.1 20 - 21 2382 64
GenBank mRNA AB002336.1 - 21 - 22 2799 64
AK096848.1 - 20 - 21 2636 64
AK126875.1 - 20 - 21 2276 64
BC013885.2 - 19 - 20 2505 64
BC040259.1 - 22 - 23 2838 64
BC113858.1 - 3 - 4 236 64
BC113899.1 - 3 - 4 235 64
BC131796.1 - 3 - 4 237 64
BX537978.1 - 11 - 12 3226 64
CR936825.1 - 18 - 19 2116 64

View Full Product Details