The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021928.3 | NP_068747.1 | 3 - 4 | 437 | 122 |
GenBank mRNA | AK026302.1 | - | 3 - 4 | 412 | 122 |
AK092634.1 | - | 2 - 3 | 1420 | 122 | |
AL136660.1 | - | 3 - 4 | 407 | 122 | |
AY359044.1 | - | 3 - 4 | 395 | 122 | |
BC047290.1 | - | 3 - 4 | 380 | 122 | |
BC053883.1 | - | 2 - 3 | 2331 | 122 | |
DB285025.1 | - | 3 - 4 | 439 | 122 |