The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142648.1 | NP_001136120.1 | 7 - 8 | 678 | 65 |
NM_020150.4 | NP_064535.1 | 6 - 7 | 608 | 65 | |
GenBank mRNA | AF086166.1 | - | 2 - 3 | 234 | 65 |
AF261717.1 | - | 6 - 7 | 575 | 65 | |
AF318066.1 | - | 5 - 6 | 475 | 65 | |
AK298591.1 | - | 5 - 6 | 441 | 65 | |
AK312469.1 | - | 6 - 7 | 563 | 65 | |
AL136724.1 | - | 6 - 7 | 591 | 65 | |
AY008268.1 | - | 6 - 7 | 599 | 65 | |
BC003658.2 | - | 6 - 7 | 573 | 65 | |
BG327663.1 | - | 7 - 8 | 646 | 65 |