Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KDM5D
Entrez Gene ID
8284
Gene Name
lysine demethylase 5D
Gene Aliases
HY, HYA, JARID1D, SMCY
Chromosome Location
Chr.Y: 19692495 - 19745347 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146705.1 NP_001140177.1 9 - 10 1380 73
NM_001146706.1 NP_001140178.1 8 - 9 1209 73
NM_004653.4 NP_004644.2 9 - 10 1380 73
XM_005262560.2 XP_005262617.1 8 - 9 1653 73
XM_005262561.2 XP_005262618.1 9 - 10 1788 73
XM_011531468.2 XP_011529770.1 9 - 10 1788 73
GenBank mRNA AK304027.1 - 9 - 10 1167 73
BC132721.1 - 9 - 10 1303 73
BC144101.1 - 9 - 10 1303 73
BC144102.1 - 9 - 10 1303 73
BC146767.1 - 8 - 9 1031 73
D87072.1 - 8 - 9 1031 73
U52191.1 - 9 - 10 1367 73
U52365.1 - 8 - 9 1122 73

View Full Product Details