The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271762.1 | NP_001258691.1 | 7 - 8 | 839 | 67 |
NM_001271763.1 | NP_001258692.1 | 6 - 7 | 814 | 67 | |
NM_017803.4 | NP_060273.1 | 8 - 9 | 919 | 67 | |
GenBank mRNA | AB101210.1 | - | 8 - 9 | 583 | 67 |
AK000406.1 | - | 7 - 8 | 486 | 67 | |
AK290578.1 | - | 8 - 9 | 621 | 67 | |
AK296663.1 | - | 6 - 7 | 466 | 67 | |
BC006527.2 | - | 8 - 9 | 562 | 67 |