The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000789.3 | NP_000780.1 | 18 - 19 | 2770 | 128 |
NM_001178057.1 | NP_001171528.1 | 7 - 8 | 1065 | 128 | |
NM_152830.2 | NP_690043.1 | 7 - 8 | 1065 | 128 | |
XM_006721737.3 | XP_006721800.2 | 7 - 8 | 1178 | 128 | |
GenBank mRNA | AB208971.1 | - | 18 - 19 | 2760 | 128 |
AK301988.1 | - | 7 - 8 | 1036 | 128 | |
AK302019.1 | - | 6 - 7 | 747 | 128 | |
J04144.1 | - | 18 - 19 | 2758 | 128 | |
JQ927553.1 | - | 18 - 19 | 2770 | 128 | |
M26657.1 | - | 7 - 8 | 1036 | 128 | |
M29981.1 | - | 7 - 8 | 1114 | 128 | |
X16295.1 | - | 7 - 8 | 1042 | 128 |