The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005157.5 | NP_005148.2 | 7 - 8 | 1460 | 73 |
NM_007313.2 | NP_009297.2 | 7 - 8 | 1764 | 73 | |
GenBank mRNA | AB209456.1 | - | 7 - 8 | 1470 | 73 |
AB209642.1 | - | 7 - 8 | 1692 | 73 | |
BC117451.1 | - | 7 - 8 | 1390 | 73 | |
FJ785401.1 | - | 4 - 5 | 502 | 73 | |
JF272500.1 | - | 4 - 5 | 560 | 73 | |
JX565024.1 | - | 4 - 5 | 611 | 73 | |
JX565025.1 | - | 4 - 5 | 611 | 73 | |
M14752.1 | - | 7 - 8 | 1632 | 73 | |
X16416.1 | - | 7 - 8 | 1415 | 73 |