Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDZD11
Entrez Gene ID
51248
Gene Name
PDZ domain containing 11
Gene Aliases
AIPP1, PDZK11, PISP
Chromosome Location
Chr.X: 70286361 - 70290040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016484.4 NP_057568.1 5 - 6 432 80
XM_011530973.2 XP_011529275.1 6 - 7 596 80
XM_017029568.1 XP_016885057.1 5 - 6 763 80
GenBank mRNA AF151061.1 - 5 - 6 402 80
AK024746.1 - 5 - 6 400 80
AK311539.1 - 4 - 5 294 80
AY358829.1 - 5 - 6 638 80
BC012996.2 - 5 - 6 371 80
BC089433.1 - 4 - 5 329 80
BG708698.1 - 5 - 6 436 80
BX537725.1 - 5 - 6 506 80
CR457149.1 - 4 - 5 330 80

View Full Product Details